Learn More
MSC SPI Class ZZ Minus Plug Gage, Steel, 0.952 in. Diameter, 0.000200 in. Tolerance, 1 ea
Catalog No. 5000515033
Encompass
This Class ZZ minus plug and pin gage is designed for precision measurement and inspection applications. Constructed from high-quality steel, it offers durability and accuracy in a variety of industrial or laboratory settings.
  • Manufactured from durable steel
  • Precision tolerance of 0.000200 in.
  • Designed for reliable hole diameter verification
  • Class ZZ rating for consistent performance
  • Made in the USA
Catalog No. 50-005-15033 Supplier MSC Supplier No. 78381480
This Class ZZ minus plug and pin gage is designed for precision measurement and inspection applications. Constructed from high-quality steel, it offers durability and accuracy in a variety of industrial or laboratory settings.
  • Manufactured from durable steel
  • Precision tolerance of 0.000200 in.
  • Designed for reliable hole diameter verification
  • Class ZZ rating for consistent performance
  • Made in the USA