Learn More
MSC SPI Class ZZ Plus Plug Gage, Steel, 0.2110 in. Diameter, 0.0002 in. Tolerance, 1 ea
Catalog No. 5000514867
Encompass
This Class ZZ Plus plug gage is designed for precision measurement and inspection tasks. Constructed from durable steel, this gage provides accurate diameter verification for metalworking and quality control applications.
  • Diameter of 0.2110 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel
Catalog No. 50-005-14867 Supplier MSC Supplier No. 78383965
This Class ZZ Plus plug gage is designed for precision measurement and inspection tasks. Constructed from durable steel, this gage provides accurate diameter verification for metalworking and quality control applications.
  • Diameter of 0.2110 in.
  • Tolerance of 0.0002 in.
  • Constructed from steel