Learn More
MSC SPI Plug & Pin Gage, 0.0420 Inch Diameter, Class ZZ Minus, Black Oxide Finish
Catalog No. 5000528069
Encompass
This SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurately checking hole sizes. With a diameter of 0.0420 inches and a minus tolerance, it ensures precise measurements for quality control and manufacturing applications. The black oxide finish provides enhanced durability and corrosion resistance.
  • Precision ground to 0.0420 inch diameter for accurate measurements.
  • Class ZZ minus tolerance ensures high-precision gauging.
  • Black oxide finish for improved wear and corrosion resistance.
  • Ideal for verifying small hole dimensions in various applications.
Catalog No. 50-005-28069 Supplier MSC Supplier No. 62256334
This SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurately checking hole sizes. With a diameter of 0.0420 inches and a minus tolerance, it ensures precise measurements for quality control and manufacturing applications. The black oxide finish provides enhanced durability and corrosion resistance.
  • Precision ground to 0.0420 inch diameter for accurate measurements.
  • Class ZZ minus tolerance ensures high-precision gauging.
  • Black oxide finish for improved wear and corrosion resistance.
  • Ideal for verifying small hole dimensions in various applications.