Learn More
MSC SPI Plug & Pin Gage, 0.2010 in Diameter, Class ZZ Minus, Steel
Catalog No. 5000528693
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.2010 inch diameter and is constructed from durable steel. It is designed for precise measurement applications, offering a tolerance of 0.000200 inches.
  • Constructed from durable steel.
  • Features a 0.2010 inch diameter.
  • Offers a precise tolerance of 0.000200 inches.
  • Designed as a Class ZZ Minus gage.
Catalog No. 50-005-28693 Supplier MSC Supplier No. 78373974
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage features a 0.2010 inch diameter and is constructed from durable steel. It is designed for precise measurement applications, offering a tolerance of 0.000200 inches.
  • Constructed from durable steel.
  • Features a 0.2010 inch diameter.
  • Offers a precise tolerance of 0.000200 inches.
  • Designed as a Class ZZ Minus gage.