Learn More
MSC SPI Plug & Pin Gage, 0.215 in, Class ZZ Minus, Black Oxide
Catalog No. 5000530206
Encompass
This plug and pin gage is designed for precise measurement in various industrial and laboratory applications. Featuring a 0.215 inch diameter, it is ideal for accurately checking bore and hole sizes with high precision. The durable construction and black oxide finish ensure reliable performance for quality control and inspection tasks.
  • Black oxide finish provides durability
  • 0.215 inch diameter for precise measurements
  • Class ZZ Minus tolerance for high accuracy
  • Ideal for checking bore and hole sizes
  • Suitable for industrial and laboratory use
Catalog No. 50-005-30206 Supplier MSC Supplier No. 84500545
This plug and pin gage is designed for precise measurement in various industrial and laboratory applications. Featuring a 0.215 inch diameter, it is ideal for accurately checking bore and hole sizes with high precision. The durable construction and black oxide finish ensure reliable performance for quality control and inspection tasks.
  • Black oxide finish provides durability
  • 0.215 inch diameter for precise measurements
  • Class ZZ Minus tolerance for high accuracy
  • Ideal for checking bore and hole sizes
  • Suitable for industrial and laboratory use