Learn More
MSC SPI Plug & Pin Gage, 0.2300 in, Class ZZ Minus, Steel
Catalog No. 5000530899
Encompass
This plug and pin gage is designed for precise measurement applications. It features a 0.2300 in diameter and is constructed from durable steel, ensuring accuracy and longevity for quality control and inspection tasks.
  • Class ZZ minus
  • Diameter of measurement 0.2300 in
  • Tolerance of 0.000200 in
  • Constructed from steel
Catalog No. 50-005-30899 Supplier MSC Supplier No. 78374261
This plug and pin gage is designed for precise measurement applications. It features a 0.2300 in diameter and is constructed from durable steel, ensuring accuracy and longevity for quality control and inspection tasks.
  • Class ZZ minus
  • Diameter of measurement 0.2300 in
  • Tolerance of 0.000200 in
  • Constructed from steel