Learn More
MSC SPI Plug & Pin Gage, 0.2560 in, Class ZZ Minus, Steel
Catalog No. 5000530309
Encompass
This plug and pin gage is designed for precise measurement and inspection. Featuring a 0.2560 in diameter and Class ZZ Minus accuracy, it ensures high precision for various applications. Constructed from durable steel, this gage provides consistent performance in demanding environments.
  • 0.2560 in diameter for accurate measurements.
  • Class ZZ Minus for high precision.
  • Made from durable steel.
  • 0.0002 in tolerance for tight specifications.
  • 2 in overall length for standard use.
Catalog No. 50-005-30309 Supplier MSC Supplier No. 78374527
This plug and pin gage is designed for precise measurement and inspection. Featuring a 0.2560 in diameter and Class ZZ Minus accuracy, it ensures high precision for various applications. Constructed from durable steel, this gage provides consistent performance in demanding environments.
  • 0.2560 in diameter for accurate measurements.
  • Class ZZ Minus for high precision.
  • Made from durable steel.
  • 0.0002 in tolerance for tight specifications.
  • 2 in overall length for standard use.