Learn More
MSC SPI Plug and Pin Gage, 0.2610 in, Class ZZ Minus, Steel
Catalog No. 5000535579
Encompass
This is a precision plug and pin gage designed for accurate measurement applications. Constructed from durable steel, it features a 0.2610 inch diameter with a Class ZZ Minus tolerance, ensuring high accuracy for critical gaging tasks.
  • 0.2610 inch diameter
  • Class ZZ Minus
  • Tolerance of 0.000200 inch
  • Made from steel
  • Decimal inch measurement system
Catalog No. 50-005-35579 Supplier MSC Supplier No. 78374576
This is a precision plug and pin gage designed for accurate measurement applications. Constructed from durable steel, it features a 0.2610 inch diameter with a Class ZZ Minus tolerance, ensuring high accuracy for critical gaging tasks.
  • 0.2610 inch diameter
  • Class ZZ Minus
  • Tolerance of 0.000200 inch
  • Made from steel
  • Decimal inch measurement system