Learn More
MSC SPI Plug and Pin Gage, 0.3960 in, Class ZZ Plus, Steel
Catalog No. 5000531666
Encompass
This product is a Class ZZ Plus Plug and Pin Gage with a diameter of 0.3960 inches. It is constructed from steel and designed for precise measurement applications, offering a tight tolerance of 0.000200 inches.
  • Class ZZ Plus tolerance
  • Suitable for plug and pin gaging
  • Constructed from steel
  • Measures 0.3960 inch diameter
  • Provides a tolerance of 0.000200 inch
Catalog No. 50-005-31666 Supplier MSC Supplier No. 78385812
This product is a Class ZZ Plus Plug and Pin Gage with a diameter of 0.3960 inches. It is constructed from steel and designed for precise measurement applications, offering a tight tolerance of 0.000200 inches.
  • Class ZZ Plus tolerance
  • Suitable for plug and pin gaging
  • Constructed from steel
  • Measures 0.3960 inch diameter
  • Provides a tolerance of 0.000200 inch