Learn More
MSC SPI Plug & Pin Gage, 0.6530 in Diameter, 0.0002 in Tolerance, Steel
Catalog No. 5000528984
Encompass
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.6530 inch diameter and is constructed from durable steel, offering a tolerance of 0.0002 inches.
  • Class ZZ minus plug and pin gage
  • 0.6530 in diameter
  • 0.0002 in tolerance
  • Made of steel
  • Manufactured in China
Catalog No. 50-005-28984 Supplier MSC Supplier No. 78378494
This plug and pin gage is designed for precise measurement and inspection tasks. It features a 0.6530 inch diameter and is constructed from durable steel, offering a tolerance of 0.0002 inches.
  • Class ZZ minus plug and pin gage
  • 0.6530 in diameter
  • 0.0002 in tolerance
  • Made of steel
  • Manufactured in China