Learn More
MSC SPI Plug & Pin Gage, 0.6690 in Diameter, Steel, Class ZZ Minus
Catalog No. 5000528710
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision tool designed for accurate measurement. It features a 0.6690 inch diameter and is made from durable steel, ensuring reliability in various applications.
  • Designed for high-precision measurement tasks.
  • Constructed from durable steel for extended tool life.
  • Features a 0.6690 inch diameter for specific gaging requirements.
  • Adheres to Class ZZ Minus specifications for tight tolerance control.
  • Provides an exact tolerance of 0.000200 "
Catalog No. 50-005-28710 Supplier MSC Supplier No. 78378650
This Class ZZ Minus Plug & Pin Gage is a precision tool designed for accurate measurement. It features a 0.6690 inch diameter and is made from durable steel, ensuring reliability in various applications.
  • Designed for high-precision measurement tasks.
  • Constructed from durable steel for extended tool life.
  • Features a 0.6690 inch diameter for specific gaging requirements.
  • Adheres to Class ZZ Minus specifications for tight tolerance control.
  • Provides an exact tolerance of 0.000200 "