Learn More
MSC SPI Plug and Pin Gage, 0.8300 in, Class ZZ Minus
Catalog No. 5000536412
Encompass
This Class ZZ Minus Plug and Pin Gage has a 0.8300 in diameter. It is constructed from steel and features a tolerance of 0.000200 in. The gage is brand new and includes a factory warranty.
  • 0.8300 in diameter for precise measurements
  • Class ZZ Minus for specific gauging requirements
  • Features a tolerance of 0.000200 in
  • Constructed from durable steel
  • Utilizes decimal inch system of measurement
  • Comes with a factory warranty
Catalog No. 50-005-36412 Supplier MSC Supplier No. 78380268
This Class ZZ Minus Plug and Pin Gage has a 0.8300 in diameter. It is constructed from steel and features a tolerance of 0.000200 in. The gage is brand new and includes a factory warranty.
  • 0.8300 in diameter for precise measurements
  • Class ZZ Minus for specific gauging requirements
  • Features a tolerance of 0.000200 in
  • Constructed from durable steel
  • Utilizes decimal inch system of measurement
  • Comes with a factory warranty