Learn More
MSC SPI Plug & Pin Gage, 0.951 in Diameter, Class ZZ Minus, 0.0002 in Tolerance, Black Oxide
Catalog No. 5000510579
Encompass
This plug and pin gage features a 0.951 inch diameter and a black oxide finish. It has a Class ZZ Minus rating and a tolerance of 0.0002 inches, making it suitable for precise measurements.
  • Minimum order quantity of 1.
  • Package quantity of 1.
Catalog No. 50-005-10579 Supplier MSC Supplier No. 85666089
This plug and pin gage features a 0.951 inch diameter and a black oxide finish. It has a Class ZZ Minus rating and a tolerance of 0.0002 inches, making it suitable for precise measurements.
  • Minimum order quantity of 1.
  • Package quantity of 1.