Learn More
MSC SPI Plug & Pin Gage, 0.976 in, 0.0002 in Tolerance, Black Oxide
Catalog No. 5000528949
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.976 inch diameter, making it suitable for precise measurement and inspection tasks. It boasts a black oxide finish for enhanced durability and a tight 0.0002 inch tolerance, ensuring high accuracy in its applications.
  • Class ZZ Minus Plug & Pin Gage type
  • 0.976 inch diameter
  • Black oxide finish for durability
  • 0.0002 inch tolerance for high precision
Catalog No. 50-005-28949 Supplier MSC Supplier No. 85666337
This Class ZZ Minus Plug & Pin Gage features a 0.976 inch diameter, making it suitable for precise measurement and inspection tasks. It boasts a black oxide finish for enhanced durability and a tight 0.0002 inch tolerance, ensuring high accuracy in its applications.
  • Class ZZ Minus Plug & Pin Gage type
  • 0.976 inch diameter
  • Black oxide finish for durability
  • 0.0002 inch tolerance for high precision