Learn More
MSC SPI Plug & Pin Gage, 0.9990 inch, Class ZZ Minus, Black Oxide
Catalog No. 5000510804
Encompass
This Plug & Pin Gage from SPI features a precise 0.9990 inch diameter and a durable black oxide finish. Designed as a Class ZZ Minus gage, it is suitable for various measurement applications requiring high accuracy.
  • Precise 0.9990 inch diameter measurement
  • Class ZZ Minus designation ensures high accuracy
  • Durable black oxide coating for enhanced longevity
  • Tolerance of 0.000200 inch
Catalog No. 50-005-10804 Supplier MSC Supplier No. 85666568
This Plug & Pin Gage from SPI features a precise 0.9990 inch diameter and a durable black oxide finish. Designed as a Class ZZ Minus gage, it is suitable for various measurement applications requiring high accuracy.
  • Precise 0.9990 inch diameter measurement
  • Class ZZ Minus designation ensures high accuracy
  • Durable black oxide coating for enhanced longevity
  • Tolerance of 0.000200 inch