Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0130 in, Black Oxide
Catalog No. 5000530217
Encompass
The SPI Plug & Pin Gage is designed for precise measurement applications. It features a a 0.0130 inch diameter and a durable black oxide finish. This Class ZZ Minus gage has a tolerance of -0.000200 "
  • Product is in stock.
  • Available for next business day delivery.
  • Backed by a hassle-free guarantee.
Catalog No. 50-005-30217 Supplier MSC Supplier No. 56243603
The SPI Plug & Pin Gage is designed for precise measurement applications. It features a a 0.0130 inch diameter and a durable black oxide finish. This Class ZZ Minus gage has a tolerance of -0.000200 "
  • Product is in stock.
  • Available for next business day delivery.
  • Backed by a hassle-free guarantee.