Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0240 in, Black Oxide
Catalog No. 5000514720
Encompass
SPI Class ZZ Minus Plug & Pin Gages are engineered for high-precision measurement. This specific gage features a 0.0240 inch diameter and a robust black oxide finish, making it ideal for verifying bore sizes or other internal dimensions with strict tolerance requirements.
  • Class ZZ Minus for very tight tolerance applications.
  • 0.0240 inch diameter ensures accurate sizing.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Essential tool for quality control and inspection processes.
Catalog No. 50-005-14720 Supplier MSC Supplier No. 56243710
SPI Class ZZ Minus Plug & Pin Gages are engineered for high-precision measurement. This specific gage features a 0.0240 inch diameter and a robust black oxide finish, making it ideal for verifying bore sizes or other internal dimensions with strict tolerance requirements.
  • Class ZZ Minus for very tight tolerance applications.
  • 0.0240 inch diameter ensures accurate sizing.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Essential tool for quality control and inspection processes.