Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0370" Dia, Black Oxide
Catalog No. 5000528800
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Class ZZ Minus for precise measurements.
  • 0.0370 inch diameter.
  • Black oxide coating for durability.
  • Includes a Certificate of Accuracy with NIST Traceability.
Catalog No. 50-005-28800 Supplier MSC Supplier No. 56243843
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Class ZZ Minus for precise measurements.
  • 0.0370 inch diameter.
  • Black oxide coating for durability.
  • Includes a Certificate of Accuracy with NIST Traceability.