Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0590 in, Black Oxide
Catalog No. 5000528784
Encompass
This gage is a Class ZZ Minus Plug & Pin Gage designed for precise measurement. It features a 0.0590 in diameter and a durable black oxide coating.
  • Class ZZ Minus
  • Diameter of measurement: 0.0590 in
  • Tolerance: -0.000200 in
  • Black oxide coating
  • Country of origin: China
Catalog No. 50-005-28784 Supplier MSC Supplier No. 56244072
This gage is a Class ZZ Minus Plug & Pin Gage designed for precise measurement. It features a 0.0590 in diameter and a durable black oxide coating.
  • Class ZZ Minus
  • Diameter of measurement: 0.0590 in
  • Tolerance: -0.000200 in
  • Black oxide coating
  • Country of origin: China