Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0820 in Diameter, Black Oxide
Catalog No. 5000531129
Encompass
This SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurately checking hole diameters. Featuring a durable black oxide finish, it is engineered to provide reliable performance in various industrial and workshop settings. The gage has a diameter of 0.0820 inches and an overall length of 2 inches, making it suitable for a wide range of inspection tasks.
  • Provides accurate measurement of hole diameters.
  • Features a durable black oxide finish for enhanced protection.
  • Designed for precision in industrial and workshop environments.
  • Suitable for a variety of inspection tasks.
Catalog No. 50-005-31129 Supplier MSC Supplier No. 56244304
This SPI Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurately checking hole diameters. Featuring a durable black oxide finish, it is engineered to provide reliable performance in various industrial and workshop settings. The gage has a diameter of 0.0820 inches and an overall length of 2 inches, making it suitable for a wide range of inspection tasks.
  • Provides accurate measurement of hole diameters.
  • Features a durable black oxide finish for enhanced protection.
  • Designed for precision in industrial and workshop environments.
  • Suitable for a variety of inspection tasks.