Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0860 in, Black Oxide
Catalog No. 5000529049
Encompass
This is a Class ZZ Minus Plug & Pin Gage manufactured by SPI, designed for precise measurement and inspection tasks. It features a specific diameter of 0.0860 inches and is finished with black oxide for enhanced durability.
  • Class ZZ Minus for accurate measurement.
  • 0.0860 inch diameter for specific applications.
  • Black oxide coating for enhanced durability and corrosion resistance.
  • Used for precise measurement and inspection.
Catalog No. 50-005-29049 Supplier MSC Supplier No. 56244346
This is a Class ZZ Minus Plug & Pin Gage manufactured by SPI, designed for precise measurement and inspection tasks. It features a specific diameter of 0.0860 inches and is finished with black oxide for enhanced durability.
  • Class ZZ Minus for accurate measurement.
  • 0.0860 inch diameter for specific applications.
  • Black oxide coating for enhanced durability and corrosion resistance.
  • Used for precise measurement and inspection.