Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0880 in Dia, Black Oxide
Catalog No. 5000514519
Encompass
This plug and pin gage is a Class ZZ Minus measurement tool, featuring a precise 0.0880 inch diameter and a durable black oxide coating. It is designed for accurate and reliable gauging in various applications.
  • Measures precisely at 0.0880 inch diameter.
  • Offers a tight tolerance of -0.000200 "
  • Features a black oxide coating for increased durability and corrosion resistance.
  • Manufactured with quality in China.
Catalog No. 50-005-14519 Supplier MSC Supplier No. 56244361
This plug and pin gage is a Class ZZ Minus measurement tool, featuring a precise 0.0880 inch diameter and a durable black oxide coating. It is designed for accurate and reliable gauging in various applications.
  • Measures precisely at 0.0880 inch diameter.
  • Offers a tight tolerance of -0.000200 "
  • Features a black oxide coating for increased durability and corrosion resistance.
  • Manufactured with quality in China.