Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.0920 inch Diameter, Black Oxide
Catalog No. 5000529050
Encompass
This plug and pin gage is engineered for precise measurement applications, featuring a specific diameter and a durable coating to ensure accuracy and longevity in industrial and scientific environments.
  • Class ZZ Minus designation
  • 0.0920 inch diameter
  • -0.000200 inch tolerance
  • Black oxide finish for durability
Catalog No. 50-005-29050 Supplier MSC Supplier No. 56244403
This plug and pin gage is engineered for precise measurement applications, featuring a specific diameter and a durable coating to ensure accuracy and longevity in industrial and scientific environments.
  • Class ZZ Minus designation
  • 0.0920 inch diameter
  • -0.000200 inch tolerance
  • Black oxide finish for durability