Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1040 inch, Black Oxide
Catalog No. 5000527671
Encompass
This plug and pin gage features a measurement diameter of 0.1040 inches. It has a tolerance of -0.000200 inches and is finished with a black oxide coating, making it suitable for precise measurement applications.
  • Class ZZ Minus plug and pin gage for accurate measurement.
  • 0.1040 inch measurement diameter.
  • Tight tolerance of -0.000200 inch for high precision.
  • Black oxide coating for durability.
Catalog No. 50-005-27671 Supplier MSC Supplier No. 56244528
This plug and pin gage features a measurement diameter of 0.1040 inches. It has a tolerance of -0.000200 inches and is finished with a black oxide coating, making it suitable for precise measurement applications.
  • Class ZZ Minus plug and pin gage for accurate measurement.
  • 0.1040 inch measurement diameter.
  • Tight tolerance of -0.000200 inch for high precision.
  • Black oxide coating for durability.