Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1090 in, Black Oxide
Catalog No. 5000529047
Encompass
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance for accurate inspections and a durable black oxide finish for enhanced protection.
  • Class ZZ Minus tolerance
  • 0.1090 in diameter
  • Tolerance of -0.000200 in
  • Black oxide finish
  • Country of origin: China
Catalog No. 50-005-29047 Supplier MSC Supplier No. 56244577
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance for accurate inspections and a durable black oxide finish for enhanced protection.
  • Class ZZ Minus tolerance
  • 0.1090 in diameter
  • Tolerance of -0.000200 in
  • Black oxide finish
  • Country of origin: China