Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1130 in Diameter, Black Oxide
Catalog No. 5000530384
Encompass
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate inspection. With a 0.1130 in diameter and a durable black oxide finish, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Diameter of measurement: 0.1130 in
  • Tolerance: -0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China
  • Includes: Certificate of accuracy with NIST traceability
Catalog No. 50-005-30384 Supplier MSC Supplier No. 56244619
The SPI Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for accurate inspection. With a 0.1130 in diameter and a durable black oxide finish, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Diameter of measurement: 0.1130 in
  • Tolerance: -0.000200 in
  • Coating/finish: Black oxide
  • Country of origin: China
  • Includes: Certificate of accuracy with NIST traceability