Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1200 in Dia, Black Oxide
Catalog No. 5000528799
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.1200 inch and a black oxide coating, making it suitable for various industrial and laboratory uses.
  • Precise measurement of 0.1200 inch diameter
  • Tight tolerance of -0.000200 inch
  • Durable black oxide coating
Catalog No. 50-005-28799 Supplier MSC Supplier No. 56244684
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a diameter of 0.1200 inch and a black oxide coating, making it suitable for various industrial and laboratory uses.
  • Precise measurement of 0.1200 inch diameter
  • Tight tolerance of -0.000200 inch
  • Durable black oxide coating