Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1380 in, Steel
Catalog No. 5000515463
Encompass
This plug and pin gage is a Class ZZ Minus gage designed for precise measurement applications. It is made from steel and has an overall length of 2 inches.
  • Features a Class ZZ Minus designation for accuracy.
  • Offers a measurement diameter of 0.1380 "
  • Maintains a tight tolerance of 0.000200 "
  • Constructed from durable steel material.
  • Suitable for setting micrometers and verniers.
  • Ideal for NoGo inspection in various measuring devices.
  • Comes with a certificate of accuracy with NIST traceability.
Catalog No. 50-005-15463 Supplier MSC Supplier No. 78373347
This plug and pin gage is a Class ZZ Minus gage designed for precise measurement applications. It is made from steel and has an overall length of 2 inches.
  • Features a Class ZZ Minus designation for accuracy.
  • Offers a measurement diameter of 0.1380 "
  • Maintains a tight tolerance of 0.000200 "
  • Constructed from durable steel material.
  • Suitable for setting micrometers and verniers.
  • Ideal for NoGo inspection in various measuring devices.
  • Comes with a certificate of accuracy with NIST traceability.