Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1490 in Diameter, Steel
Catalog No. 5000531514
Encompass
This plug and pin gage is a Class ZZ Minus gage with a 0.1490 inch diameter. It is constructed from durable steel, making it suitable for precise measurement applications.
  • Class ZZ Minus for accurate gaging
  • Diameter of 0.1490 in
  • Tolerance of 0.000200 in
  • Constructed from steel for durability
  • Manufactured in China
Catalog No. 50-005-31514 Supplier MSC Supplier No. 78373453
This plug and pin gage is a Class ZZ Minus gage with a 0.1490 inch diameter. It is constructed from durable steel, making it suitable for precise measurement applications.
  • Class ZZ Minus for accurate gaging
  • Diameter of 0.1490 in
  • Tolerance of 0.000200 in
  • Constructed from steel for durability
  • Manufactured in China