Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1660 in, Steel
Catalog No. 5000528970
Encompass
This Class ZZ Minus Plug & Pin Gage is constructed from steel and features a precise 0.1660 in diameter with a tolerance of 0.000200 in, making it suitable for accurate measurement applications.
Catalog No. 50-005-28970 Supplier MSC Supplier No. 78373628
This Class ZZ Minus Plug & Pin Gage is constructed from steel and features a precise 0.1660 in diameter with a tolerance of 0.000200 in, making it suitable for accurate measurement applications.