Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1730 in, Steel, Black Oxide
Catalog No. 5000528872
Encompass
This plug and pin gage is a Class ZZ Minus tool, designed for precise measurements. It features a 0.1730 inch diameter and a durable black oxide finish.
  • Class ZZ Minus for accurate gaging.
  • 0.1730 inch diameter for precise measurements.
  • Constructed from steel for durability.
  • Black oxide coating for enhanced protection.
  • 2 inch overall length.
  • Tolerance of -0.000200 "
Catalog No. 50-005-28872 Supplier MSC Supplier No. 56245228
Add to Cart
Add to Cart
This plug and pin gage is a Class ZZ Minus tool, designed for precise measurements. It features a 0.1730 inch diameter and a durable black oxide finish.
  • Class ZZ Minus for accurate gaging.
  • 0.1730 inch diameter for precise measurements.
  • Constructed from steel for durability.
  • Black oxide coating for enhanced protection.
  • 2 inch overall length.
  • Tolerance of -0.000200 "