Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1820 in, Steel
Catalog No. 5000531517
Encompass
This Class ZZ Minus Plug and Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.1820 inch diameter and a tight tolerance for accurate gaging.
  • Precise measurement for accurate gaging
  • 0.1820 inch diameter
  • Steel construction for durability
  • 0.000200 inch tolerance
Catalog No. 50-005-31517 Supplier MSC Supplier No. 78373784
Add to Cart
Add to Cart
This Class ZZ Minus Plug and Pin Gage is designed for precise measurement applications. Made from durable steel, it features a 0.1820 inch diameter and a tight tolerance for accurate gaging.
  • Precise measurement for accurate gaging
  • 0.1820 inch diameter
  • Steel construction for durability
  • 0.000200 inch tolerance