Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1830 in Dia, Black Oxide
Catalog No. 5000527782
Encompass
SPI Plug & Pin Gages are precision measurement tools designed for accurate inspection. This Class ZZ Minus gage features a 0.1830 inch diameter and a durable black oxide finish, making it suitable for a variety of quality control applications.
  • Precision measurement tool
  • Ideal for inspection and quality control
  • Features a 0.1830 in diameter
  • Class ZZ Minus tolerance for precise fits
  • Durable black oxide finish
Catalog No. 50-005-27782 Supplier MSC Supplier No. 56245327
Add to Cart
Add to Cart
SPI Plug & Pin Gages are precision measurement tools designed for accurate inspection. This Class ZZ Minus gage features a 0.1830 inch diameter and a durable black oxide finish, making it suitable for a variety of quality control applications.
  • Precision measurement tool
  • Ideal for inspection and quality control
  • Features a 0.1830 in diameter
  • Class ZZ Minus tolerance for precise fits
  • Durable black oxide finish