Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.1980 Dia, Steel
Catalog No. 5000528690
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for accurate measurement applications. It features a precise diameter and tight tolerance, making it suitable for checking bore sizes and other dimensional inspections.
  • Diameter of measurement: 0.1980 in
  • Tolerance: 0.000200 in (0.0002 mm)
  • Material: Steel
  • Country of origin: China
Catalog No. 50-005-28690 Supplier MSC Supplier No. 78373941
This Class ZZ Minus Plug & Pin Gage is designed for accurate measurement applications. It features a precise diameter and tight tolerance, making it suitable for checking bore sizes and other dimensional inspections.
  • Diameter of measurement: 0.1980 in
  • Tolerance: 0.000200 in (0.0002 mm)
  • Material: Steel
  • Country of origin: China