Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.2010 inch Diameter, Black Oxide
Catalog No. 5000530438
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a durable black oxide coating for enhanced longevity and resistance, making it suitable for industrial and laboratory environments requiring accurate dimensional verification.
  • Class ZZ minus tolerance
  • Diameter of measurement 0.2010 inch
  • Black oxide coating and finish
  • Tolerance of -0.000200 inch
Catalog No. 50-005-30438 Supplier MSC Supplier No. 56245509
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a durable black oxide coating for enhanced longevity and resistance, making it suitable for industrial and laboratory environments requiring accurate dimensional verification.
  • Class ZZ minus tolerance
  • Diameter of measurement 0.2010 inch
  • Black oxide coating and finish
  • Tolerance of -0.000200 inch