Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.2170 in Diameter, Steel
Catalog No. 5000528809
Encompass
This plug and pin gage is a precision measuring instrument designed for accurate inspection. It features a 0.2170 in diameter and is constructed from durable steel, ensuring reliability and longevity. With a tight tolerance of 0.000200 in, it provides precise measurements for quality control applications.
  • Precisely measures 0.2170 in diameter.
  • Constructed from durable steel.
  • Features a high tolerance of 0.000200 in.
Catalog No. 50-005-28809 Supplier MSC Supplier No. 78374139
This plug and pin gage is a precision measuring instrument designed for accurate inspection. It features a 0.2170 in diameter and is constructed from durable steel, ensuring reliability and longevity. With a tight tolerance of 0.000200 in, it provides precise measurements for quality control applications.
  • Precisely measures 0.2170 in diameter.
  • Constructed from durable steel.
  • Features a high tolerance of 0.000200 in.