Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.2510 inch, Steel
Catalog No. 5000528694
Encompass
This precision plug and pin gage is designed for accurate measurement. Made from durable steel, it features a Class ZZ Minus tolerance and a 0.2510 inch diameter, making it suitable for a variety of calibration and inspection tasks.
  • Precision plug and pin gage
  • Made from durable steel
  • Class ZZ minus tolerance
  • 0.2510 inch diameter
  • 0.000200 inch tolerance
  • Suitable for calibration and inspection applications
Catalog No. 50-005-28694 Supplier MSC Supplier No. 78374477
This precision plug and pin gage is designed for accurate measurement. Made from durable steel, it features a Class ZZ Minus tolerance and a 0.2510 inch diameter, making it suitable for a variety of calibration and inspection tasks.
  • Precision plug and pin gage
  • Made from durable steel
  • Class ZZ minus tolerance
  • 0.2510 inch diameter
  • 0.000200 inch tolerance
  • Suitable for calibration and inspection applications