Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.2820 Dia, Steel
Catalog No. 5000531515
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, featuring a 0.2820 inch diameter. Manufactured from durable steel, it ensures accuracy with a tolerance of 0.000200 inch (0.0002 mm).
  • Diameter of measurement: 0.2820 inch
  • Class: ZZ Minus
  • Tolerance: 0.000200 inch (0.0002 mm)
  • Material: Steel
Catalog No. 50-005-31515 Supplier MSC Supplier No. 78374782
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications, featuring a 0.2820 inch diameter. Manufactured from durable steel, it ensures accuracy with a tolerance of 0.000200 inch (0.0002 mm).
  • Diameter of measurement: 0.2820 inch
  • Class: ZZ Minus
  • Tolerance: 0.000200 inch (0.0002 mm)
  • Material: Steel