Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.2890" Dia, Black Oxide
Catalog No. 5000527986
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a nominal diameter of 0.2890 inches. Its black oxide finish provides durability and corrosion resistance. This gage is ideal for quality control, inspection, and calibration applications requiring high accuracy.
  • Designed for precise measurement.
  • Features a nominal diameter of 0.2890 inches.
  • Offers a tolerance of -0.000200 inches.
  • Coated with black oxide for enhanced durability.
  • Suitable for various inspection and calibration tasks.
Catalog No. 50-005-27986 Supplier MSC Supplier No. 56246382
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a nominal diameter of 0.2890 inches. Its black oxide finish provides durability and corrosion resistance. This gage is ideal for quality control, inspection, and calibration applications requiring high accuracy.
  • Designed for precise measurement.
  • Features a nominal diameter of 0.2890 inches.
  • Offers a tolerance of -0.000200 inches.
  • Coated with black oxide for enhanced durability.
  • Suitable for various inspection and calibration tasks.