Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.3190 in, Steel
Catalog No. 5000530308
Encompass
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a measurement diameter of 0.3190 inches, making it suitable for checking bore sizes and hole locations. Constructed from durable steel, this gage ensures reliability and accuracy in various industrial settings.
  • Class ZZ Minus tolerance
  • 0.3190 in diameter of measurement
  • 0.000200 in tolerance
  • Steel material construction
Catalog No. 50-005-30308 Supplier MSC Supplier No. 78375151
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a measurement diameter of 0.3190 inches, making it suitable for checking bore sizes and hole locations. Constructed from durable steel, this gage ensures reliability and accuracy in various industrial settings.
  • Class ZZ Minus tolerance
  • 0.3190 in diameter of measurement
  • 0.000200 in tolerance
  • Steel material construction