Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.3510 inch Dia, Black Oxide
Catalog No. 5000531820
Encompass
This is a Class ZZ Minus Plug & Pin Gage with a 0.3510 inch diameter. It features a black oxide coating, which helps protect against corrosion and wear.
  • Diameter of measurement: 0.3510 inch
  • Tolerance: -0.000200 inch
  • Coating and finish: Black oxide
Catalog No. 50-005-31820 Supplier MSC Supplier No. 56247000
This is a Class ZZ Minus Plug & Pin Gage with a 0.3510 inch diameter. It features a black oxide coating, which helps protect against corrosion and wear.
  • Diameter of measurement: 0.3510 inch
  • Tolerance: -0.000200 inch
  • Coating and finish: Black oxide