Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.352 Dia, Black Oxide
Catalog No. 5000530076
Encompass
This plug and pin gage is designed for precise measurement and inspection. It features a 0.352 inch diameter and a black oxide finish, providing durability and clear identification for quality control applications.
  • Class ZZ Minus tolerance
  • Black oxide coating
  • 0.352 inch diameter
  • Tolerance of -0.000200 inches
Catalog No. 50-005-30076 Supplier MSC Supplier No. 56247018
This plug and pin gage is designed for precise measurement and inspection. It features a 0.352 inch diameter and a black oxide finish, providing durability and clear identification for quality control applications.
  • Class ZZ Minus tolerance
  • Black oxide coating
  • 0.352 inch diameter
  • Tolerance of -0.000200 inches