Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.3570 inch, Black Oxide
Catalog No. 5000531809
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.3570 inch diameter and a durable black oxide finish, making it suitable for various industrial and laboratory settings where accurate gaging is required.
  • Designed for precise measurement.
  • Features a 0.3570 inch diameter.
  • Equipped with a black oxide finish for durability.
Catalog No. 50-005-31809 Supplier MSC Supplier No. 56247067
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 0.3570 inch diameter and a durable black oxide finish, making it suitable for various industrial and laboratory settings where accurate gaging is required.
  • Designed for precise measurement.
  • Features a 0.3570 inch diameter.
  • Equipped with a black oxide finish for durability.