Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.3640 in Diameter, Steel
Catalog No. 5000531516
Encompass
This plug & pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a specific diameter for accurate gauging, constructed from durable steel for longevity.
  • Class ZZ Minus for precise measurement
  • Diameter of 0.3640 in
  • Made from durable steel
  • Tolerance of 0.000200 in
Catalog No. 50-005-31516 Supplier MSC Supplier No. 78375607
This plug & pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance and a specific diameter for accurate gauging, constructed from durable steel for longevity.
  • Class ZZ Minus for precise measurement
  • Diameter of 0.3640 in
  • Made from durable steel
  • Tolerance of 0.000200 in