Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4080 in, Black Oxide
Catalog No. 5000527988
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.4080 inch diameter and a durable black oxide coating. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for critical tasks. The overall length is 2 inches.
  • Designed for precise measurement applications.
  • Features a black oxide coating for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers and NoGo inspection.
  • Suitable for use with verniers and other measuring devices.
Catalog No. 50-005-27988 Supplier MSC Supplier No. 56247570
This Class ZZ Minus Plug & Pin Gage features a 0.4080 inch diameter and a durable black oxide coating. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for critical tasks. The overall length is 2 inches.
  • Designed for precise measurement applications.
  • Features a black oxide coating for durability.
  • Includes a certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers and NoGo inspection.
  • Suitable for use with verniers and other measuring devices.