Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4120 in Diameter, Steel
Catalog No. 5000516366
Encompass
This Class ZZ Minus plug and pin gage is designed for precise measurement applications. It features a nominal diameter of 0.4120 inches and is constructed from durable steel, ensuring accuracy and reliability for quality control tasks.
  • Precisely measures 0.4120 in diameter.
  • Features a tight tolerance of 0.000200 in for accuracy.
  • Constructed from steel for durability and longevity.
  • Manufactured to Class ZZ Minus specifications.
  • Made in China.
Catalog No. 50-005-16366 Supplier MSC Supplier No. 78376084
This Class ZZ Minus plug and pin gage is designed for precise measurement applications. It features a nominal diameter of 0.4120 inches and is constructed from durable steel, ensuring accuracy and reliability for quality control tasks.
  • Precisely measures 0.4120 in diameter.
  • Features a tight tolerance of 0.000200 in for accuracy.
  • Constructed from steel for durability and longevity.
  • Manufactured to Class ZZ Minus specifications.
  • Made in China.