Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4300 in, Steel
Catalog No. 5000530821
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a diameter of 0.4300 in and a tolerance of 0.0002 in, making it suitable for tasks requiring high accuracy.
  • Provides precise measurement
  • Suitable for various precision measurement tasks, including gunsmithing
  • Class ZZ Minus type
  • 0.4300 in diameter
  • Tolerance of 0.0002 in
  • Made from steel
Catalog No. 50-005-30821 Supplier MSC Supplier No. 78376266
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Constructed from durable steel, it features a diameter of 0.4300 in and a tolerance of 0.0002 in, making it suitable for tasks requiring high accuracy.
  • Provides precise measurement
  • Suitable for various precision measurement tasks, including gunsmithing
  • Class ZZ Minus type
  • 0.4300 in diameter
  • Tolerance of 0.0002 in
  • Made from steel