Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4320 inch Diameter, Steel
Catalog No. 5000528807
Encompass
This plug and pin gage is designed for precision measurement tasks. It features a Class ZZ Minus tolerance and a specific diameter, making it suitable for verifying bore sizes and other critical dimensions in various applications.
  • Provides precise measurement with a 0.4320 inch diameter.
  • Features a tolerance of 0.000200 inch for accurate gauging.
  • Constructed from durable steel for long-lasting use.
  • Class ZZ Minus designation ensures high accuracy for critical applications.
Catalog No. 50-005-28807 Supplier MSC Supplier No. 78376282
This plug and pin gage is designed for precision measurement tasks. It features a Class ZZ Minus tolerance and a specific diameter, making it suitable for verifying bore sizes and other critical dimensions in various applications.
  • Provides precise measurement with a 0.4320 inch diameter.
  • Features a tolerance of 0.000200 inch for accurate gauging.
  • Constructed from durable steel for long-lasting use.
  • Class ZZ Minus designation ensures high accuracy for critical applications.