Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4380 in Dia, Steel
Catalog No. 5000528689
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a diameter of 0.4380 inches. Constructed from steel, it offers a tight tolerance of 0.000200 inch, making it suitable for demanding applications requiring high accuracy.
  • Class ZZ Minus
  • Diameter of 0.4380 inch
  • Tolerance of 0.000200 inch
  • Constructed from steel
Catalog No. 50-005-28689 Supplier MSC Supplier No. 78376340
Add to Cart
Add to Cart
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement with a diameter of 0.4380 inches. Constructed from steel, it offers a tight tolerance of 0.000200 inch, making it suitable for demanding applications requiring high accuracy.
  • Class ZZ Minus
  • Diameter of 0.4380 inch
  • Tolerance of 0.000200 inch
  • Constructed from steel