Learn More
MSC SPI Plug & Pin Gage, Class ZZ Minus, 0.4430 in Diameter, Steel
Catalog No. 5000528715
Encompass
This Class ZZ Minus Plug & Pin Gage has a diameter of 0.4430 in and is made from steel. The gage features a tight tolerance of 0.000200 in, ensuring precision measurements for various applications.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter: 0.4430 in
  • Class: ZZ Minus
  • Tolerance of 0.000200 in
  • Material: Steel
  • Country of origin: Various
Catalog No. 50-005-28715 Supplier MSC Supplier No. 78376399
This Class ZZ Minus Plug & Pin Gage has a diameter of 0.4430 in and is made from steel. The gage features a tight tolerance of 0.000200 in, ensuring precision measurements for various applications.
  • Class ZZ Minus Plug & Pin Gage
  • Diameter: 0.4430 in
  • Class: ZZ Minus
  • Tolerance of 0.000200 in
  • Material: Steel
  • Country of origin: Various